siRNA Details
| Gene Symbol: Hyal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207599.2 | NP_997482.2 | 3 | 1056 | ||
| GenBank mRNA | AB100602.1 | - | 2 | 920 | ||
| BC097259.1 | - | 3 | 1056 | 7453545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hyal3| Gene Symbol: Hyal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207599.2 | NP_997482.2 | 3 | 1056 | ||
| GenBank mRNA | AB100602.1 | - | 2 | 920 | ||
| BC097259.1 | - | 3 | 1056 | 7453545 | ||