siRNA Details
| Gene Symbol: Hyal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207599.2 | NP_997482.2 | 2 | 387 | ||
| XM_008766504.1 | XP_008764726.1 | 2 | 386 | |||
| GenBank mRNA | AB100602.1 | - | 1 | 251 | ||
| BC097259.1 | - | 2 | 387 | 7453545 | ||

Hyal3
Genomic Map