siRNA Details
| Gene Symbol: Ifrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047871.1 | NP_001041336.1 | 7, 8 | 995 | ||
| GenBank mRNA | BC161810.1 | - | 7, 8 | 929 | 9023096 | |
| DQ480751.1 | - | 7, 8 | 995 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifrd2| Gene Symbol: Ifrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047871.1 | NP_001041336.1 | 7, 8 | 995 | ||
| GenBank mRNA | BC161810.1 | - | 7, 8 | 929 | 9023096 | |
| DQ480751.1 | - | 7, 8 | 995 | |||