siRNA Details
| Gene Symbol: Rbm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100548.1 | NP_001094018.1 | 17 | 1701 | ||
| XM_006243738.2 | XP_006243800.1 | 17 | 1955 | |||
| XM_006243739.2 | XP_006243801.1 | 17 | 1593 | |||
| GenBank mRNA | BC166477.1 | - | 17 | 1701 | 9102173 | |

Rbm5
Genomic Map