siRNA Details
| Gene Symbol: Uba7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106856.1 | NP_001100326.1 | 8 | 986 | ||
| XM_006243741.2 | XP_006243803.1 | 8 | 938 | |||
| XM_006243742.2 | XP_006243804.1 | 8 | 938 | |||
| XM_006243743.2 | XP_006243805.1 | 8 | 938 | |||
| XM_008766505.1 | XP_008764727.1 | 7 | 634 | |||

Uba7
Genomic Map