siRNA Details
| Gene Symbol: P4htm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_217285.8 | XP_217285.6 | 9 | 2022 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

P4htm| Gene Symbol: P4htm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_217285.8 | XP_217285.6 | 9 | 2022 | ||