siRNA Details
| Gene Symbol: Spink8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106860.1 | NP_001100330.1 | 4 | 334 | ||
| XM_008766512.1 | XP_008764734.1 | 5 | 499 | |||
| GenBank mRNA | BC166531.1 | - | 3 | 1662 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spink8| Gene Symbol: Spink8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106860.1 | NP_001100330.1 | 4 | 334 | ||
| XM_008766512.1 | XP_008764734.1 | 5 | 499 | |||
| GenBank mRNA | BC166531.1 | - | 3 | 1662 | ||