siRNA Details
| Gene Symbol: Smarcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243770.2 | XP_006243832.1 | 14 | 1389 | ||
| XM_006243771.2 | XP_006243833.1 | 14 | 1389 | |||
| XM_006243772.2 | XP_006243834.1 | 14 | 1389 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smarcc1| Gene Symbol: Smarcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243770.2 | XP_006243832.1 | 14 | 1389 | ||
| XM_006243771.2 | XP_006243833.1 | 14 | 1389 | |||
| XM_006243772.2 | XP_006243834.1 | 14 | 1389 | |||