siRNA Details
| Gene Symbol: Ltf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106864.1 | NP_001100334.1 | 14 | 1708 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ltf| Gene Symbol: Ltf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106864.1 | NP_001100334.1 | 14 | 1708 | ||