siRNA Details
| Gene Symbol: Susd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766656.1 | XP_008764878.1 | 4 | 1372 | ||
| XM_236674.7 | XP_236674.5 | 5 | 1561 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Susd5| Gene Symbol: Susd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766656.1 | XP_008764878.1 | 4 | 1372 | ||
| XM_236674.7 | XP_236674.5 | 5 | 1561 | |||