siRNA Details
| Gene Symbol: Cmtm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198754.2 | NP_942049.1 | 1 | 481 | ||
| GenBank mRNA | AF538323.1 | - | 1 | 284 | ||
| BC088094.1 | - | 1 | 481 | 7325894 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmtm8| Gene Symbol: Cmtm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198754.2 | NP_942049.1 | 1 | 481 | ||
| GenBank mRNA | AF538323.1 | - | 1 | 284 | ||
| BC088094.1 | - | 1 | 481 | 7325894 | ||