siRNA Details
| Gene Symbol: Exog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106866.1 | NP_001100336.1 | 4, 5 | 567 | ||
| XM_006244088.2 | XP_006244150.1 | 5, 6 | 628 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exog| Gene Symbol: Exog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106866.1 | NP_001100336.1 | 4, 5 | 567 | ||
| XM_006244088.2 | XP_006244150.1 | 5, 6 | 628 | |||