siRNA Details
| Gene Symbol: PTP4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003463.4 | NP_003454.1 | 4 | 1398 | ||
| XM_011536111.1 | XP_011534413.1 | 4 | 737 | |||
| XM_011536112.1 | XP_011534414.1 | 4 | 1496 | |||
| XM_017011270.1 | XP_016866759.1 | 4 | 754 | |||
| XM_017011271.1 | XP_016866760.1 | 4 | 734 | |||
| XM_017011272.1 | XP_016866761.1 | 4 | 1398 | |||
| GenBank mRNA | AB451314.1 | - | 3 | 245 | ||
| AB451444.1 | - | 3 | 245 | |||
| AK312526.1 | - | 4 | 882 | |||
| BC023975.2 | - | 4 | 840 | 2960001 | ||
| BC045571.1 | - | 4 | 1398 | 4826233 | ||
| CR749458.1 | - | 6 | 1232 | |||
| U48296.1 | - | 3 | 637 | |||

PTP4A1
Genomic Map