siRNA Details
| Gene Symbol: Zfp445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191802.1 | NP_001178731.1 | 7 | 2590 | ||
| XM_006244181.1 | XP_006244243.1 | 7 | 2664 | |||
| XM_006244183.2 | XP_006244245.1 | 7 | 2591 | |||
| GenBank mRNA | FQ226616.1 | - | 6 | 2337 | ||

Zfp445
Genomic Map