siRNA Details
| Gene Symbol: RGD1311745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008329.1 | NP_001008330.1 | 3 | 448 | ||
| GenBank mRNA | BC085913.1 | - | 3 | 448 | 7106823 | |
| FQ226096.1 | - | 3 | 480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311745| Gene Symbol: RGD1311745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008329.1 | NP_001008330.1 | 3 | 448 | ||
| GenBank mRNA | BC085913.1 | - | 3 | 448 | 7106823 | |
| FQ226096.1 | - | 3 | 480 | |||