siRNA Details
| Gene Symbol: Zdhhc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039014.1 | NP_001034103.1 | 5, 6 | 775 | ||
| XM_006244188.2 | XP_006244250.1 | 6, 7 | 948 | |||
| GenBank mRNA | AY886522.1 | - | 5, 6 | 775 | ||
| BC128697.1 | - | 6, 7 | 894 | 7444822 | ||

Zdhhc3
Genomic Map