siRNA Details
| Gene Symbol: Vwa5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198755.1 | NP_942050.1 | 2132 | |||
| XM_006235502.2 | XP_006235564.1 | 17 | 2261 | |||
| XM_006235503.2 | XP_006235565.1 | 16 | 2198 | |||
| XM_006235504.2 | XP_006235566.1 | 16 | 2180 | |||
| XM_006242839.2 | XP_006242901.1 | 17 | 2261 | |||
| XM_006242840.2 | XP_006242902.1 | 16 | 2198 | |||
| XM_006242841.2 | XP_006242903.1 | 16 | 2180 | |||
| GenBank mRNA | AB121446.1 | - | 2132 | |||

Vwa5a
Genomic Map