siRNA Details
| Gene Symbol: RGD1559995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_235194.3 | XP_235194.1 | 1 | 296 | ||
| Gene Symbol: Dppa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_236761.3 | XP_236761.1 | 2 | 916 | ||
| Gene Symbol: RGD1564306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_229957.4 | XP_229957.2 | 1 | 318 | ||

RGD1559995
Genomic Map