siRNA Details
| Gene Symbol: Trim42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013955.1 | NP_001013977.1 | 1 | 442 | ||
| XM_008766513.1 | XP_008764735.1 | 1 | 481 | |||
| GenBank mRNA | BC087151.1 | - | 1 | 442 | 7131982 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim42| Gene Symbol: Trim42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013955.1 | NP_001013977.1 | 1 | 442 | ||
| XM_008766513.1 | XP_008764735.1 | 1 | 481 | |||
| GenBank mRNA | BC087151.1 | - | 1 | 442 | 7131982 | |