siRNA Details
| Gene Symbol: Dus3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034923.1 | NP_001030095.1 | 11 | 1609 | ||
| XM_006244314.2 | XP_006244376.1 | 11 | 1764 | |||
| GenBank mRNA | BC105780.1 | - | 11 | 1609 | 7932299 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dus3l| Gene Symbol: Dus3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034923.1 | NP_001030095.1 | 11 | 1609 | ||
| XM_006244314.2 | XP_006244376.1 | 11 | 1764 | |||
| GenBank mRNA | BC105780.1 | - | 11 | 1609 | 7932299 | |