siRNA Details
| Gene Symbol: Ndufa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212517.1 | NP_997682.1 | 1, 2 | 154 | ||
| GenBank mRNA | AY272059.1 | - | 1, 2 | 154 | ||
| FQ209810.1 | - | 1, 2 | 134 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufa11| Gene Symbol: Ndufa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212517.1 | NP_997682.1 | 1, 2 | 154 | ||
| GenBank mRNA | AY272059.1 | - | 1, 2 | 154 | ||
| FQ209810.1 | - | 1, 2 | 134 | |||