siRNA Details
Gene Symbol: Znrf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024878.1 | NP_001020049.1 | 1 | 825 | ||
GenBank mRNA | BC098025.1 | - | 1 | 825 | 7455918 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Znrf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024878.1 | NP_001020049.1 | 1 | 825 | ||
GenBank mRNA | BC098025.1 | - | 1 | 825 | 7455918 |