siRNA Details
| Gene Symbol: Efhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106879.1 | NP_001100349.1 | 10 | 1998 | ||
| GenBank mRNA | BC166482.1 | - | 10 | 1999 | 7098775 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efhb| Gene Symbol: Efhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106879.1 | NP_001100349.1 | 10 | 1998 | ||
| GenBank mRNA | BC166482.1 | - | 10 | 1999 | 7098775 | |