siRNA Details
| Gene Symbol: Unc5cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106882.1 | NP_001100352.1 | 4 | 1044 | ||
| XM_006244421.2 | XP_006244483.1 | 6 | 1875 | |||
| XM_006244422.2 | XP_006244484.1 | 5 | 1441 | |||
| XM_006244423.2 | XP_006244485.1 | 4 | 1603 | |||
| XM_008766843.1 | XP_008765065.1 | 4 | 1603 | |||

Unc5cl
Genomic Map