siRNA Details
| Gene Symbol: Trem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106885.1 | NP_001100355.1 | 2 | 424 | ||
| XM_006244426.2 | XP_006244488.1 | 569 | ||||
| XM_006244427.2 | XP_006244489.1 | 571 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trem1| Gene Symbol: Trem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106885.1 | NP_001100355.1 | 2 | 424 | ||
| XM_006244426.2 | XP_006244488.1 | 569 | ||||
| XM_006244427.2 | XP_006244489.1 | 571 | ||||