siRNA Details
| Gene Symbol: RGD1561662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244500.2 | XP_006244562.1 | 4 | 2036 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561662| Gene Symbol: RGD1561662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244500.2 | XP_006244562.1 | 4 | 2036 | ||