siRNA Details
| Gene Symbol: Guca1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106887.1 | NP_001100357.1 | 3 | 559 | ||
| XM_006244428.2 | XP_006244490.1 | 2 | 514 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Guca1a| Gene Symbol: Guca1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106887.1 | NP_001100357.1 | 3 | 559 | ||
| XM_006244428.2 | XP_006244490.1 | 2 | 514 | |||