siRNA Details
| Gene Symbol: Mrpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034136.1 | NP_001029308.1 | 3, 4 | 428 | ||
| GenBank mRNA | BC100081.1 | - | 3, 4 | 428 | 7325700 | |
| FQ214924.1 | - | 3, 4 | 461 | |||
| FQ229593.1 | - | 3, 4 | 462 | |||

Mrpl2
Genomic Map