siRNA Details
| Gene Symbol: Tmem151b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191804.1 | NP_001178733.1 | 2 | 286 | ||
| XM_008766851.1 | XP_008765073.1 | 1 | 2277 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem151b| Gene Symbol: Tmem151b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191804.1 | NP_001178733.1 | 2 | 286 | ||
| XM_008766851.1 | XP_008765073.1 | 1 | 2277 | |||