siRNA Details
| Gene Symbol: Aars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106891.1 | NP_001100361.1 | 6 | 961 | ||
| XM_006244513.2 | XP_006244575.1 | 6 | 1966 | |||
| XM_006244515.2 | XP_006244577.1 | 6 | 1962 | |||
| GenBank mRNA | BC166719.1 | - | 6 | 950 | ||

Aars2
Genomic Map