siRNA Details
| Gene Symbol: Enpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106892.1 | NP_001100362.1 | 2 | 461 | ||
| XM_006244598.2 | XP_006244660.1 | 3 | 1857 | |||
| XM_006244600.2 | XP_006244662.1 | 2 | 1742 | |||
| XM_006244602.2 | XP_006244664.1 | 2 | 443 | |||
| XM_008766852.1 | XP_008765074.1 | 3 | 557 | |||
| XM_008766853.1 | XP_008765075.1 | 2 | 351 | |||

Enpp4
Genomic Map