siRNA Details
| Gene Symbol: Cyp39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106893.1 | NP_001100363.1 | 5, 6 | 1047 | ||
| XM_006244604.2 | XP_006244666.1 | 5, 6 | 1047 | |||
| XM_006244605.1 | XP_006244667.1 | 5, 6 | 1047 | |||
| XM_008766854.1 | XP_008765076.1 | 5, 6 | 930 | |||
| XM_008766855.1 | XP_008765077.1 | 5, 6 | 928 | |||

Cyp39a1
Genomic Map