siRNA Details
| Gene Symbol: Pla2g7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009353.1 | NP_001009353.1 | 8 | 929 | ||
| XM_006244606.2 | XP_006244668.1 | 8 | 1046 | |||
| XM_008766856.1 | XP_008765078.1 | 7 | 2689 | |||
| GenBank mRNA | BC088457.1 | - | 8 | 929 | 7127300 | |

Pla2g7
Genomic Map