siRNA Details
Gene Symbol: Il17f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015011.1 | NP_001015011.1 | 4 | 343 | ||
GenBank mRNA | BC091568.1 | - | 4 | 343 | 7443410 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il17f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015011.1 | NP_001015011.1 | 4 | 343 | ||
GenBank mRNA | BC091568.1 | - | 4 | 343 | 7443410 |