siRNA Details
| Gene Symbol: RGD1309049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013956.1 | NP_001013978.1 | 1 | 390 | ||
| GenBank mRNA | BC082106.1 | - | 1 | 390 | 7134835 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1309049| Gene Symbol: RGD1309049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013956.1 | NP_001013978.1 | 1 | 390 | ||
| GenBank mRNA | BC082106.1 | - | 1 | 390 | 7134835 | |