siRNA Details
| Gene Symbol: Prim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024762.1 | NP_001019933.1 | 13 | 1371 | ||
| XM_006244686.2 | XP_006244748.1 | 11 | 1200 | |||
| XM_006244689.2 | XP_006244751.1 | 11 | 1151 | |||
| XM_006244690.2 | XP_006244752.1 | 11 | 1217 | |||
| XM_006244691.1 | XP_006244753.1 | 11 | 1218 | |||
| GenBank mRNA | AJ011607.1 | - | 10 | 1055 | ||
| BC097454.1 | - | 13 | 1371 | 7456950 | ||

Prim2
Genomic Map