siRNA Details
| Gene Symbol: Ptpn18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013111.1 | NP_001013129.1 | 6, 7 | 481 | ||
| GenBank mRNA | BC093398.1 | - | 5, 6 | 582 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpn18| Gene Symbol: Ptpn18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013111.1 | NP_001013129.1 | 6, 7 | 481 | ||
| GenBank mRNA | BC093398.1 | - | 5, 6 | 582 | ||