siRNA Details
| Gene Symbol: Plekhb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106899.1 | NP_001100369.1 | 8 | 672 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhb2| Gene Symbol: Plekhb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106899.1 | NP_001100369.1 | 8 | 672 | ||