siRNA Details
| Gene Symbol: Mettl21c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106908.1 | NP_001100378.1 | 3 | 298 | ||
| XM_008767022.1 | XP_008765244.1 | 2 | 1090 | |||
| GenBank mRNA | FQ215023.1 | - | 267 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl21c| Gene Symbol: Mettl21c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106908.1 | NP_001100378.1 | 3 | 298 | ||
| XM_008767022.1 | XP_008765244.1 | 2 | 1090 | |||
| GenBank mRNA | FQ215023.1 | - | 267 | |||