siRNA Details
| Gene Symbol: Tex30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106909.1 | NP_001100379.1 | 4 | 396 | ||
| GenBank mRNA | FQ230447.1 | - | 3 | 288 | ||
| FQ230685.1 | - | 3 | 288 | |||
| FQ232543.1 | - | 3 | 288 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex30| Gene Symbol: Tex30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106909.1 | NP_001100379.1 | 4 | 396 | ||
| GenBank mRNA | FQ230447.1 | - | 3 | 288 | ||
| FQ230685.1 | - | 3 | 288 | |||
| FQ232543.1 | - | 3 | 288 | |||