siRNA Details
| Gene Symbol: Ankrd44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191807.3 | NP_001178736.3 | 2244 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd44| Gene Symbol: Ankrd44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191807.3 | NP_001178736.3 | 2244 | |||