siRNA Details
| Gene Symbol: MGC94335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004251.1 | NP_001004251.1 | 3 | 960 | ||
| GenBank mRNA | BC079246.1 | - | 3 | 960 | 7132862 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC94335| Gene Symbol: MGC94335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004251.1 | NP_001004251.1 | 3 | 960 | ||
| GenBank mRNA | BC079246.1 | - | 3 | 960 | 7132862 | |