siRNA Details
| Gene Symbol: Tyw5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170473.1 | NP_001163944.1 | 8 | 1042 | ||
| XM_006244910.2 | XP_006244972.1 | 7 | 648 | |||
| GenBank mRNA | BC158619.1 | - | 8 | 1023 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tyw5| Gene Symbol: Tyw5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170473.1 | NP_001163944.1 | 8 | 1042 | ||
| XM_006244910.2 | XP_006244972.1 | 7 | 648 | |||
| GenBank mRNA | BC158619.1 | - | 8 | 1023 | ||