siRNA Details
| Gene Symbol: Orc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012003.1 | NP_001012003.1 | 6, 7 | 505 | ||
| XM_006244956.2 | XP_006245018.1 | 7, 8 | 791 | |||
| GenBank mRNA | AB164704.1 | - | 6, 7 | 505 | ||
| BC129076.1 | - | 6, 7 | 639 | |||

Orc2
Genomic Map