siRNA Details
| Gene Symbol: Nif3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024763.1 | NP_001019934.1 | 3 | 802 | ||
| XM_006244958.2 | XP_006245020.1 | 5 | 943 | |||
| XM_006244960.2 | XP_006245022.1 | 5 | 914 | |||
| XM_006244961.2 | XP_006245023.1 | 5 | 939 | |||
| XM_006244962.2 | XP_006245024.1 | 4 | 812 | |||
| XM_006244963.2 | XP_006245025.1 | 5 | 946 | |||
| XM_008767082.1 | XP_008765304.1 | 5 | 947 | |||
| GenBank mRNA | BC097437.1 | - | 3 | 802 | 7455562 | |
| BC097992.1 | - | 5 | 875 | 7446502 | ||

Nif3l1
Genomic Map