siRNA Details
| Gene Symbol: Nif3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024763.1 | NP_001019934.1 | 5 | 1039 | ||
| XM_006244963.2 | XP_006245025.1 | 7 | 1183 | |||
| XM_008767082.1 | XP_008765304.1 | 7 | 1184 | |||
| GenBank mRNA | BC097437.1 | - | 5 | 1039 | 7455562 | |

Nif3l1
Genomic Map