siRNA Details
| Gene Symbol: Ppil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175707.3 | NP_783638.1 | 4 | 467 | ||
| XM_006244964.2 | XP_006245026.1 | 4 | 537 | |||
| GenBank mRNA | AF315802.1 | - | 4 | 255 | ||
| BC087645.1 | - | 4 | 467 | 7311008 | ||
| FQ224392.1 | - | 4 | 264 | |||

Ppil3
Genomic Map