siRNA Details
| Gene Symbol: Fzd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767173.1 | XP_008765395.1 | 2 | 768 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fzd7| Gene Symbol: Fzd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767173.1 | XP_008765395.1 | 2 | 768 | ||