siRNA Details
| Gene Symbol: Carf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106915.1 | NP_001100385.1 | 5 | 534 | ||
| XM_006244969.1 | XP_006245031.1 | 3 | 463 | |||
| XM_006244970.2 | XP_006245032.1 | 4 | 749 | |||
| XM_006244971.1 | XP_006245033.1 | 2 | 304 | |||
| XM_006244974.2 | XP_006245036.1 | 2 | 989 | |||

Carf
Genomic Map