siRNA Details
Gene Symbol: Carf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106915.1 | NP_001100385.1 | 12 | 1258 | ||
XM_006244969.1 | XP_006245031.1 | 9 | 1120 | |||
XM_006244970.2 | XP_006245032.1 | 10 | 1406 | |||
XM_006244971.1 | XP_006245033.1 | 8 | 961 | |||
XM_006244973.2 | XP_006245035.1 | 7 | 682 | |||
XM_006244974.2 | XP_006245036.1 | 8 | 1646 |
